siRNA Details
| Gene Symbol: SPTSSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138288.3 | NP_612145.2 | 2 | 237 | ||
| GenBank mRNA | AK001993.1 | - | 2 | 218 | ||
| AK315409.1 | - | 2 | 215 | |||
| BC016805.1 | - | 2 | 219 | 4075637 | ||
| BC016808.1 | - | 2 | 219 | 4077762 | ||
| BC021701.1 | - | 2 | 218 | 4069530 | ||
| BC068480.1 | - | 2 | 331 | 5294549 | ||

SPTSSA
Genomic Map