siRNA Details
| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 2199 | ||
| GenBank mRNA | AK126379.1 | - | 1 | 2139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM122A| Gene Symbol: FAM122A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138333.4 | NP_612206.4 | 1 | 2199 | ||
| GenBank mRNA | AK126379.1 | - | 1 | 2139 | ||