siRNA Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099286.2 | NP_001092756.1 | 6 | 959 | ||
| NM_001318738.1 | NP_001305667.1 | 7 | 771 | |||
| NM_138419.4 | NP_612428.2 | 6 | 671 | |||
| XM_011535410.2 | XP_011533712.1 | 7 | 1023 | |||
| XM_011535412.2 | XP_011533714.1 | 5 | 764 | |||
| XM_011535413.2 | XP_011533715.1 | 5 | 3878 | |||
| GenBank mRNA | AK125758.1 | - | 7 | 728 | ||
| AK292303.1 | - | 6 | 910 | |||
| BC011716.2 | - | 3 | 294 | |||
| BC063688.1 | - | 6 | 624 | 4455541 | ||

MTFR2
Genomic Map