siRNA Details
| Gene Symbol: GINM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138785.3 | NP_620140.1 | 1, 2 | 154 | ||
| GenBank mRNA | AK289402.1 | - | 1, 2 | 141 | ||
| AK295690.1 | - | 1, 2 | 180 | |||
| AK308844.1 | - | 1, 2 | 141 | |||
| AK315729.1 | - | 1, 2 | 151 | |||
| AY358952.1 | - | 1, 2 | 272 | |||
| BC014320.1 | - | 1, 2 | 158 | 3686673 | ||

GINM1
Genomic Map