siRNA Details
| Gene Symbol: TTC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288781.1 | NP_001275710.1 | 5 | 526 | ||
| NM_001288782.1 | NP_001275711.1 | 5 | 526 | |||
| NM_001288783.1 | NP_001275712.1 | 5 | 526 | |||
| NM_144596.3 | NP_653197.2 | 6 | 556 | |||
| NM_198309.3 | NP_938051.1 | 5 | 526 | |||
| NM_198310.3 | NP_938052.1 | 5 | 526 | |||
| XM_006720035.1 | XP_006720098.1 | 4 | 808 | |||
| XM_006720037.2 | XP_006720100.1 | 4 | 808 | |||
| XM_011536432.1 | XP_011534734.1 | 4 | 808 | |||
| XM_011536433.1 | XP_011534735.1 | 4 | 808 | |||
| XM_011536434.1 | XP_011534736.1 | 4 | 808 | |||
| XM_011536435.2 | XP_011534737.1 | 3 | 212 | |||
| XM_017020977.1 | XP_016876466.1 | 4 | 808 | |||
| XM_017020978.1 | XP_016876467.1 | 4 | 808 | |||
| GenBank mRNA | AK093891.1 | - | 4 | 404 | ||
| AK124675.1 | - | 5 | 406 | |||
| AK296077.1 | - | 4 | 387 | |||
| AK315772.1 | - | 4 | 411 | |||
| AL833901.1 | - | 4 | 350 | |||
| AY366523.1 | - | 4 | 382 | |||
| AY366524.1 | - | 4 | 382 | |||
| AY373972.1 | - | 4 | 382 | |||
| BC026351.1 | - | 4 | 382 | 4799330 | ||
| BC095433.1 | - | 4 | 364 | 30368216 | ||
| BG330023.1 | - | 403 | ||||
| BX161472.1 | - | 4 | 366 | |||
| BX247959.1 | - | 6 | 680 | |||
| BX248248.1 | - | 2 | 58 | |||
| KU178849.1 | - | 4 | 330 | |||
| KU178850.1 | - | 4 | 330 | |||
| KU178851.1 | - | 4 | 330 | |||
| KU178853.1 | - | 4 | 330 | |||
| KU178854.1 | - | 4 | 330 | |||

TTC8
Genomic Map