siRNA Details
| Gene Symbol: CMTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144601.3 | NP_653202.1 | 6 | 1972 | ||
| NM_181553.2 | NP_853531.1 | 5 | 1808 | |||
| NR_037613.1 | 6 | 2192 | ||||
| NR_037619.1 | 5 | 1865 | ||||
| XM_006721131.1 | XP_006721194.1 | 6 | 2016 | |||
| XM_011522861.1 | XP_011521163.1 | 4 | 1645 | |||
| GenBank mRNA | AF479813.1 | - | 6 | 1946 | ||
| AK056324.1 | - | 6 | 2192 | |||
| AK075075.1 | - | 4 | 1505 | |||
| BC047701.1 | - | 5 | 1808 | 5770132 | ||

CMTM3
Genomic Map