siRNA Details
| Gene Symbol: RFTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144629.2 | NP_653230.2 | 2 | 674 | ||
| XM_011510595.2 | XP_011508897.1 | 2 | 864 | |||
| XM_011510596.2 | XP_011508898.1 | 2 | 864 | |||
| XM_011510597.2 | XP_011508899.1 | 2 | 864 | |||
| XM_011510598.2 | XP_011508900.1 | 2 | 864 | |||
| XM_017003332.1 | XP_016858821.1 | 2 | 864 | |||
| GenBank mRNA | AK055136.1 | - | 2 | 674 | ||
| BC093966.1 | - | 2 | 321 | 7939811 | ||
| BC111069.1 | - | 2 | 806 | 5764096 | ||
| BC113361.1 | - | 2 | 321 | 8322413 | ||

RFTN2
Genomic Map