siRNA Details
| Gene Symbol: DOCK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144658.3 | NP_653259.3 | 7 | 671 | ||
| XM_005262368.3 | XP_005262425.1 | 7 | 916 | |||
| XM_005262369.4 | XP_005262426.1 | 7 | 610 | |||
| XM_011531276.2 | XP_011529578.1 | 7 | 551 | |||
| GenBank mRNA | AK124735.1 | - | 1 | 56 | ||
| AY692226.1 | - | 7 | 671 | |||

DOCK11
Genomic Map