siRNA Details
| Gene Symbol: MARCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002266.2 | NP_001002266.1 | 7 | 1767 | ||
| NM_001282866.1 | NP_001269795.1 | 8 | 2613 | |||
| NM_145021.5 | NP_659458.2 | 7 | 2255 | |||
| XM_005271804.2 | XP_005271861.1 | 8 | 3102 | |||
| XM_006717704.2 | XP_006717767.1 | 8 | 2517 | |||
| XM_011539492.2 | XP_011537794.1 | 9 | 3153 | |||
| XM_011539493.2 | XP_011537795.1 | 10 | 3394 | |||
| XM_011539494.2 | XP_011537796.1 | 9 | 3343 | |||
| XM_011539495.1 | XP_011537797.1 | 7 | 1651 | |||
| XM_017015894.1 | XP_016871383.1 | 8 | 2307 | |||
| GenBank mRNA | AK023636.1 | - | 3 | 2056 | ||
| AK057289.1 | - | 7 | 1961 | |||
| AL833316.1 | - | 4 | 1998 | |||
| BC025394.2 | - | 7 | 1744 | 4830278 | ||
| BC066988.1 | - | 7 | 2256 | 5267214 | ||

MARCH8
Genomic Map