siRNA Details
| Gene Symbol: ZNF501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 2728 | ||
| NM_145044.3 | NP_659481.2 | 3 | 2435 | |||
| GenBank mRNA | BC013762.1 | - | 3 | 2411 | 4521303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF501| Gene Symbol: ZNF501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 2728 | ||
| NM_145044.3 | NP_659481.2 | 3 | 2435 | |||
| GenBank mRNA | BC013762.1 | - | 3 | 2411 | 4521303 | |