siRNA Details
| Gene Symbol: WDR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006615.2 | NP_001006616.1 | 5 | 596 | ||
| NM_001012361.3 | NP_001012361.1 | 6 | 729 | |||
| NM_145241.4 | NP_660284.1 | 6 | 726 | |||
| NR_134677.1 | 6 | 658 | ||||
| XM_011518194.2 | XP_011516496.1 | 5 | 640 | |||
| GenBank mRNA | AK074891.1 | - | 5 | 510 | ||
| AK093240.1 | - | 6 | 658 | |||
| AK313843.1 | - | 6 | 668 | |||
| AL833635.1 | - | 6 | 575 | |||
| BC012352.1 | - | 6 | 658 | 4634445 | ||

WDR31
Genomic Map