siRNA Details
| Gene Symbol: WDR31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006615.2 | NP_001006616.1 | 10 | 1807 | ||
| NM_001012361.3 | NP_001012361.1 | 11 | 1940 | |||
| NM_145241.4 | NP_660284.1 | 11 | 1937 | |||
| NR_134677.1 | 11 | 1908 | ||||
| XM_011518194.2 | XP_011516496.1 | 10 | 1851 | |||
| GenBank mRNA | AK074891.1 | - | 10 | 1721 | ||
| AK093240.1 | - | 11 | 1869 | |||
| AL833635.1 | - | 11 | 1825 | |||
| BC012352.1 | - | 11 | 1869 | 4634445 | ||
| BC022982.1 | - | 11 | 1866 | |||

WDR31
Genomic Map