siRNA Details
| Gene Symbol: THBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003247.3 | NP_003238.2 | 23 | 4019 | ||
| GenBank mRNA | AK292429.1 | - | 23 | 3976 | ||
| BC146676.1 | - | 22 | 3826 | 40148868 | ||
| BC150175.1 | - | 22 | 3830 | 40147958 | ||
| BX641023.1 | - | 9 | 1474 | |||
| L12350.1 | - | 23 | 3938 | |||
| M81339.1 | - | 12 | 2023 | |||

THBS2
Genomic Map