siRNA Details
| Gene Symbol: STOML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144033.1 | NP_001137505.1 | 4 | 405 | ||
| NM_145286.2 | NP_660329.1 | 3 | 321 | |||
| GenBank mRNA | AK304159.1 | - | 4 | 405 | ||
| AK313632.1 | - | 3 | 293 | |||
| BC025760.1 | - | 3 | 308 | 5204252 | ||
| DB200356.1 | - | 4 | 384 | |||

STOML3
Genomic Map