siRNA Details
| Gene Symbol: TMEM217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145316.3 | NP_660359.2 | 3 | 751 | ||
| NR_104442.1 | 2 | 653 | ||||
| XM_011514362.1 | XP_011512664.1 | 4 | 763 | |||
| XM_011514363.2 | XP_011512665.1 | 4 | 763 | |||
| XM_011514364.2 | XP_011512666.1 | 4 | 1547 | |||
| XM_011514365.2 | XP_011512667.1 | 4 | 1332 | |||
| XM_011514366.2 | XP_011512668.1 | 3 | 737 | |||
| GenBank mRNA | AK098666.1 | - | 3 | 710 | ||
| DB458508.1 | - | 2 | 135 | |||

TMEM217
Genomic Map