siRNA Details
| Gene Symbol: THBS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306212.1 | NP_001293141.1 | 14 | 1676 | ||
| NM_001306213.1 | NP_001293142.1 | 14 | 1728 | |||
| NM_001306214.1 | NP_001293143.1 | 13 | 1670 | |||
| NM_003248.5 | NP_003239.2 | 13 | 1842 | |||
| XM_017009798.1 | XP_016865287.1 | 13 | 1822 | |||
| XM_017009799.1 | XP_016865288.1 | 13 | 1822 | |||
| XM_017009800.1 | XP_016865289.1 | 13 | 1822 | |||
| GenBank mRNA | AK302842.1 | - | 13 | 1670 | ||
| AK313587.1 | - | 13 | 1658 | |||
| BC050456.1 | - | 14 | 1926 | 6191808 | ||
| Z19585.1 | - | 13 | 1630 | |||

THBS4
Genomic Map