siRNA Details
| Gene Symbol: MTPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145808.3 | NP_665807.1 | 4 | 1838 | ||
| GenBank mRNA | AK055660.1 | - | 1 | 389 | ||
| AK056225.1 | - | 4 | 1741 | |||
| AL834231.1 | - | 4 | 1710 | |||
| Gene Symbol: LUZP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128619.2 | NP_001122091.2 | 4 | 1838 | ||

MTPN
Genomic Map