siRNA Details
| Gene Symbol: PIK3AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152309.2 | NP_689522.2 | 17 | 4664 | ||
| XM_005269498.1 | XP_005269555.1 | 16 | 4187 | |||
| XM_005269499.1 | XP_005269556.1 | 16 | 4211 | |||
| XM_011539248.1 | XP_011537550.1 | 16 | 4537 | |||
| XM_011539249.1 | XP_011537551.1 | 16 | 4500 | |||

PIK3AP1
Genomic Map