siRNA Details
| Gene Symbol: FAM210A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098801.1 | NP_001092271.1 | 5 | 1491 | ||
| NM_152352.3 | NP_689565.2 | 4 | 1351 | |||
| GenBank mRNA | AK055618.1 | - | 5 | 1447 | ||
| AK309506.1 | - | 2 | 738 | |||
| AK309819.1 | - | 2 | 625 | |||
| CB987819.1 | - | 1 | 187 | |||

FAM210A
Genomic Map