siRNA Details
| Gene Symbol: ZNF440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152357.2 | NP_689570.2 | 4 | 2974 | ||
| XM_005259731.3 | XP_005259788.1 | 4 | 3034 | |||
| XM_017026253.1 | XP_016881742.1 | 3 | 3359 | |||
| XM_017026254.1 | XP_016881743.1 | 3 | 2852 | |||
| GenBank mRNA | AK092559.1 | - | 1 | 1070 | ||
| AK095252.1 | - | 4 | 2974 | |||
| BC016371.1 | - | 1 | 1071 | |||
| BC035760.1 | - | 4 | 2957 | 5556302 | ||

ZNF440
Genomic Map