siRNA Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253829.1 | NP_001240758.1 | 6 | 941 | ||
| NM_001253830.1 | NP_001240759.1 | 7 | 1002 | |||
| NM_152422.4 | NP_689635.3 | 6 | 935 | |||
| NM_177995.2 | NP_818931.1 | 7 | 1017 | |||
| XM_005251711.4 | XP_005251768.1 | 7 | 2959 | |||
| XM_011518240.2 | XP_011516542.1 | 7 | 767 | |||
| XM_011518241.2 | XP_011516543.1 | 7 | 721 | |||
| XM_017014280.1 | XP_016869769.1 | 8 | 826 | |||
| XM_017014281.1 | XP_016869770.1 | 7 | 745 | |||
| XM_017014282.1 | XP_016869771.1 | 4 | 848 | |||
| GenBank mRNA | AK124912.1 | - | 6 | 821 | ||
| AK289692.1 | - | 6 | 892 | |||
| AK310403.1 | - | 6 | 899 | |||
| AY171233.1 | - | 7 | 1022 | |||
| BC067120.1 | - | 7 | 1002 | 5539182 | ||
| DA312531.1 | - | 5 | 517 | |||

PTPDC1
Genomic Map