siRNA Details
| Gene Symbol: RNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286398.2 | NP_001273327.1 | 3 | 1240 | ||
| NM_152553.4 | NP_689766.1 | 5 | 706 | |||
| NR_104440.2 | 4 | 597 | ||||
| NR_136734.1 | 4 | 1361 | ||||
| XM_011535494.2 | XP_011533796.1 | 3 | 1347 | |||
| XM_011535496.2 | XP_011533798.1 | 2 | 1121 | |||
| XM_011535500.2 | XP_011533802.1 | 3 | 475 | |||
| GenBank mRNA | AK093417.1 | - | 4 | 597 | ||
| AK122811.1 | - | 3 | 1069 | |||
| BC026087.2 | - | 5 | 702 | 4827114 | ||
| DB238637.1 | - | 4 | 574 | |||

RNF217
Genomic Map