siRNA Details
| Gene Symbol: RNF217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286398.2 | NP_001273327.1 | 4 | 1319 | ||
| NM_152553.4 | NP_689766.1 | 6 | 785 | |||
| NR_104440.2 | 5 | 676 | ||||
| NR_136734.1 | 5 | 1440 | ||||
| XM_011535494.2 | XP_011533796.1 | 4 | 1426 | |||
| XM_011535496.2 | XP_011533798.1 | 3 | 1200 | |||
| XM_011535500.2 | XP_011533802.1 | 4 | 554 | |||
| GenBank mRNA | AK057943.1 | - | 1 | 45 | ||
| AK093417.1 | - | 5 | 676 | |||
| AK122811.1 | - | 4 | 1148 | |||
| BC026087.2 | - | 6 | 781 | 4827114 | ||

RNF217
Genomic Map