siRNA Details
| Gene Symbol: TTC39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168339.1 | NP_001161811.1 | 4 | 488 | ||
| NM_001168340.1 | NP_001161812.1 | 4 | 488 | |||
| NM_001168342.1 | NP_001161814.1 | 2 | 363 | |||
| NM_152574.2 | NP_689787.2 | 4 | 488 | |||
| XM_011517732.2 | XP_011516034.1 | 3 | 285 | |||
| XM_017014310.1 | XP_016869799.1 | 6 | 851 | |||
| XM_017014311.1 | XP_016869800.1 | 5 | 783 | |||
| XM_017014312.1 | XP_016869801.1 | 3 | 353 | |||
| GenBank mRNA | AK091187.1 | - | 4 | 374 | ||
| AK093459.1 | - | 4 | 555 | |||
| AK298998.1 | - | 4 | 460 | |||
| AK301185.1 | - | 4 | 339 | |||
| AK301427.1 | - | 2 | 279 | |||
| AK310271.1 | - | 5 | 346 | |||
| AK310482.1 | - | 4 | 488 | |||
| BC042417.1 | - | 3 | 355 | |||
| BC142985.1 | - | 4 | 467 | 8860428 | ||
| DT217817.1 | - | 3 | 460 | |||

TTC39B
Genomic Map