siRNA Details
| Gene Symbol: TRIM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152616.4 | NP_689829.3 | 2, 3 | 1250 | ||
| XM_011512740.2 | XP_011511042.1 | 2, 3 | 1245 | |||
| GenBank mRNA | AF521868.1 | - | 2, 3 | 1062 | ||
| AK097416.1 | - | 2, 3 | 1237 | |||
| AK310536.1 | - | 2, 3 | 1229 | |||
| BC130472.1 | - | 2, 3 | 1160 | 40146503 | ||
| BC130474.1 | - | 2, 3 | 1160 | 40146505 | ||

TRIM42
Genomic Map