siRNA Details
| Gene Symbol: ZNF366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152625.2 | NP_689838.1 | 5 | 2581 | ||
| GenBank mRNA | AK097115.1 | - | 5 | 2581 | ||
| BC121053.1 | - | 5 | 2452 | 40118323 | ||
| BC121054.1 | - | 5 | 2452 | 40118325 | ||
| BX647592.1 | - | 5 | 2471 | |||

ZNF366
Genomic Map