siRNA Details
Gene Symbol: THAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152658.2 | NP_689871.1 | 4 | 1652 | ||
XM_005258623.2 | XP_005258680.1 | 4 | 1461 | |||
XM_011526604.2 | XP_011524906.1 | 4 | 1661 | |||
GenBank mRNA | AK057453.1 | - | 5 | 1505 | ||
AK093048.1 | - | 4 | 1653 | |||
AK296640.1 | - | 4 | 1146 | |||
AK297630.1 | - | 3 | 962 | |||
BC046207.1 | - | 4 | 1201 | 6176484 | ||
BC072416.1 | - | 4 | 1292 | 6169558 | ||
JQ410992.1 | - | 4 | 1529 |
