siRNA Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152658.2 | NP_689871.1 | 4 | 1688 | ||
| XM_005258623.2 | XP_005258680.1 | 4 | 1497 | |||
| XM_011526604.2 | XP_011524906.1 | 4 | 1697 | |||
| GenBank mRNA | AK057453.1 | - | 5 | 1541 | ||
| AK093048.1 | - | 4 | 1689 | |||
| AK296640.1 | - | 4 | 1182 | |||
| BC046207.1 | - | 4 | 1237 | 6176484 | ||
| BC072416.1 | - | 4 | 1328 | 6169558 | ||
| JQ410992.1 | - | 4 | 1565 | |||

THAP8
Genomic Map