siRNA Details
| Gene Symbol: FBXO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142958.1 | NP_001136430.1 | 4 | 614 | ||
| NM_152676.2 | NP_689889.1 | 4 | 642 | |||
| XM_011525856.1 | XP_011524158.1 | 4 | 583 | |||
| XM_011525858.2 | XP_011524160.1 | 4 | 583 | |||
| XM_011525859.1 | XP_011524161.1 | 4 | 583 | |||
| XM_011525860.1 | XP_011524162.1 | 4 | 583 | |||
| XM_017025603.1 | XP_016881092.1 | 5 | 624 | |||
| XM_017025604.1 | XP_016881093.1 | 4 | 2652 | |||
| GenBank mRNA | AK094215.1 | - | 4 | 554 | ||
| BC029579.1 | - | 4 | 592 | 5268841 | ||
| BI829755.1 | - | 4 | 707 | |||

FBXO15
Genomic Map