siRNA Details
| Gene Symbol: FBXO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142958.1 | NP_001136430.1 | 10 | 1369 | ||
| NM_152676.2 | NP_689889.1 | 10 | 1397 | |||
| XM_011525856.1 | XP_011524158.1 | 9 | 1128 | |||
| XM_017025603.1 | XP_016881092.1 | 11 | 1379 | |||
| XM_017025604.1 | XP_016881093.1 | 10 | 3407 | |||
| GenBank mRNA | AK093252.1 | - | 4 | 2853 | ||
| AK094215.1 | - | 10 | 1309 | |||
| BC029579.1 | - | 10 | 1347 | 5268841 | ||

FBXO15
Genomic Map