siRNA Details
| Gene Symbol: STOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130161.2 | NP_001123633.1 | 3 | 2530 | ||
| NM_152709.4 | NP_689922.3 | 3 | 2530 | |||
| XM_011539454.2 | XP_011537756.1 | 3 | 2261 | |||
| GenBank mRNA | AK057864.1 | - | 1 | 149 | ||
| AK057891.1 | - | 1 | 920 | |||
| AK124681.1 | - | 3 | 2225 | |||
| AK127878.1 | - | 1 | 1571 | |||
| AY842014.1 | - | 3 | 2443 | |||
| AY842017.1 | - | 3 | 2443 | |||
| BC063627.1 | - | 1 | 911 | |||

STOX1
Genomic Map