siRNA Details
| Gene Symbol: CPNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152727.5 | NP_689940.3 | 10 | 1164 | ||
| GenBank mRNA | AF492484.1 | - | 10 | 1115 | ||
| AK094867.1 | - | 1 | 122 | |||
| AK292269.1 | - | 9 | 1073 | |||
| AK315394.1 | - | 10 | 1067 | |||
| BC053658.1 | - | 11 | 1238 | 5750192 | ||
| CR749617.1 | - | 9 | 825 | |||

CPNE2
Genomic Map