siRNA Details
| Gene Symbol: BTBD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099272.1 | NP_001092742.1 | 5 | 1254 | ||
| NM_001172418.1 | NP_001165889.1 | 4 | 1039 | |||
| NM_052893.1 | NP_443125.1 | 6 | 1454 | |||
| NM_152733.2 | NP_689946.2 | 4 | 991 | |||
| XM_011514279.2 | XP_011512581.1 | 7 | 1851 | |||
| XM_011514280.2 | XP_011512582.1 | 5 | 1532 | |||
| XM_011514281.2 | XP_011512583.1 | 5 | 1532 | |||
| XM_011514282.2 | XP_011512584.1 | 5 | 1532 | |||
| XM_017010227.1 | XP_016865716.1 | 6 | 1732 | |||
| GenBank mRNA | AB067467.1 | - | 5 | 1120 | ||
| AK057507.1 | - | 4 | 991 | |||
| BC101354.2 | - | 4 | 912 | 40025568 | ||
| BC101355.1 | - | 4 | 912 | 40025569 | ||
| BC101356.2 | - | 4 | 912 | |||
| BC101357.1 | - | 4 | 960 | 40025572 | ||
| BX537534.1 | - | 5 | 987 | |||

BTBD9
Genomic Map