siRNA Details
| Gene Symbol: C9orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278629.1 | NP_001265558.1 | 3 | 585 | ||
| NM_001278630.1 | NP_001265559.1 | 3 | 442 | |||
| NM_152786.2 | NP_689999.1 | 3 | 658 | |||
| XM_011518477.1 | XP_011516779.1 | 3 | 585 | |||
| XM_011518478.1 | XP_011516780.1 | 3 | 585 | |||
| GenBank mRNA | AK301872.1 | - | 3 | 442 | ||
| AK313867.1 | - | 3 | 574 | |||
| BC026884.1 | - | 3 | 624 | 3455891 | ||
| BC033037.2 | - | 3 | 543 | 4827930 | ||

C9orf43
Genomic Map