siRNA Details
| Gene Symbol: C9orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278629.1 | NP_001265558.1 | 13 | 1538 | ||
| NM_001278630.1 | NP_001265559.1 | 13 | 1395 | |||
| NM_152786.2 | NP_689999.1 | 13 | 1611 | |||
| XM_005251885.3 | XP_005251942.1 | 11 | 917 | |||
| XM_011518478.1 | XP_011516780.1 | 13 | 1548 | |||
| XM_011518479.2 | XP_011516781.1 | 11 | 1193 | |||
| GenBank mRNA | AK313867.1 | - | 13 | 1527 | ||
| BC026884.1 | - | 13 | 1577 | 3455891 | ||
| BC033037.2 | - | 13 | 1496 | 4827930 | ||

C9orf43
Genomic Map