siRNA Details
| Gene Symbol: SPATA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166271.2 | NP_001159743.1 | 9, 10 | 3414 | ||
| NM_001286792.1 | NP_001273721.1 | 11, 12 | 3738 | |||
| NM_001286793.1 | NP_001273722.1 | 5, 6 | 962 | |||
| NM_001286794.1 | NP_001273723.1 | 7, 8 | 1356 | |||
| NM_001286795.1 | NP_001273724.1 | 6, 7 | 1148 | |||
| NM_153023.3 | NP_694568.1 | 8, 9 | 1650 | |||
| GenBank mRNA | AK055770.1 | - | 8, 9 | 1618 | ||
| AK097217.1 | - | 2, 3 | 291 | |||
| AK123031.1 | - | 6, 7 | 1048 | |||
| AK160371.1 | - | 5, 6 | 837 | |||
| AK297394.1 | - | 5, 6 | 932 | |||
| AK298717.1 | - | 7, 8 | 1258 | |||
| AK299981.1 | - | 6, 7 | 1091 | |||
| BC109290.1 | - | 7, 8 | 1360 | 40034357 | ||
| BC109291.1 | - | 7, 8 | 1360 | 40034359 | ||
| BX648244.1 | - | 11, 12 | 3560 | |||

SPATA13
Genomic Map