siRNA Details
| Gene Symbol: C6orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005248850.3 | XP_005248907.1 | 1 | 1877 | ||
| XM_005248852.2 | XP_005248909.1 | 2 | 1088 | |||
| XM_006714997.2 | XP_006715060.1 | 2 | 1672 | |||
| XM_011514303.2 | XP_011512605.1 | 1 | 1877 | |||
| XM_017010294.1 | XP_016865783.1 | 1 | 1877 | |||
| XM_017010295.1 | XP_016865784.1 | 1 | 1877 | |||
| XM_017010296.1 | XP_016865785.1 | 2 | 1672 | |||
| XM_017010297.1 | XP_016865786.1 | 2 | 1672 | |||
| XM_017010298.1 | XP_016865787.1 | 1 | 1877 | |||
| GenBank mRNA | AK054918.1 | - | 2 | 1054 | ||

C6orf141
Genomic Map