siRNA Details
Gene Symbol: NLRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153447.4 | NP_703148.4 | 3 | 474 | ||
GenBank mRNA | AY054986.1 | - | 3 | 474 | ||
AY154460.1 | - | 3 | 474 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NLRP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153447.4 | NP_703148.4 | 3 | 474 | ||
GenBank mRNA | AY054986.1 | - | 3 | 474 | ||
AY154460.1 | - | 3 | 474 |