siRNA Details
| Gene Symbol: SAXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287049.1 | NP_001273978.1 | 3 | 424 | ||
| NM_153707.3 | NP_714918.2 | 3 | 666 | |||
| XM_011517744.1 | XP_011516046.1 | 4 | 746 | |||
| XM_011517745.1 | XP_011516047.1 | 5 | 681 | |||
| XM_011517746.2 | XP_011516048.1 | 5 | 602 | |||
| XM_011517747.2 | XP_011516049.1 | 2 | 262 | |||
| XM_017014314.1 | XP_016869803.1 | 5 | 992 | |||
| XM_017014315.1 | XP_016869804.1 | 3 | 379 | |||
| GenBank mRNA | AK292245.1 | - | 3 | 617 | ||
| BC033489.1 | - | 3 | 594 | 5170510 | ||
| BM563230.1 | - | 3 | 443 | |||

SAXO1
Genomic Map