siRNA Details
| Gene Symbol: ADGRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153834.3 | NP_722576.3 | 5 | 590 | ||
| XM_011531269.2 | XP_011529571.1 | 4 | 596 | |||
| XM_011531271.2 | XP_011529573.1 | 2 | 328 | |||
| GenBank mRNA | AY140954.1 | - | 1 | 110 | ||
| AY882585.1 | - | 5 | 590 | |||
More Information
Set Membership: |
|

ADGRG4
Genomic Map