siRNA Details
Gene Symbol: WFDC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172005.1 | NP_742002.1 | 4 | 1132 | ||
GenBank mRNA | AF454505.1 | - | 4 | 1132 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WFDC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172005.1 | NP_742002.1 | 4 | 1132 | ||
GenBank mRNA | AF454505.1 | - | 4 | 1132 |