siRNA Details
| Gene Symbol: KLHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172193.2 | NP_751943.1 | 9 | 829 | ||
| XM_011536422.2 | XP_011534724.1 | 9 | 752 | |||
| GenBank mRNA | AF111806.1 | - | 9 | 767 | ||
| AK127202.1 | - | 9 | 752 | |||
| AL833437.1 | - | 4 | 259 | |||
| BC031270.1 | - | 9 | 792 | |||
| BC101595.1 | - | 9 | 829 | 8069101 | ||
| BC101597.1 | - | 9 | 829 | 8069103 | ||

KLHDC1
Genomic Map