siRNA Details
Gene Symbol: FBXO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258211.1 | NP_001245140.1 | 8 | 1219 | ||
NM_172366.3 | NP_758954.1 | 9 | 1255 | |||
GenBank mRNA | AF453435.1 | - | 9 | 1164 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FBXO16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258211.1 | NP_001245140.1 | 8 | 1219 | ||
NM_172366.3 | NP_758954.1 | 9 | 1255 | |||
GenBank mRNA | AF453435.1 | - | 9 | 1164 |