siRNA Details
| Gene Symbol: SPRR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173080.2 | NP_775103.1 | 2 | 195 | ||
| XM_017000482.1 | XP_016855971.1 | 3 | 891 | |||
| GenBank mRNA | BC069445.1 | - | 2 | 183 | 7262727 | |
| BC069549.1 | - | 2 | 183 | 7262739 | ||
| BC112170.1 | - | 2 | 195 | 8327638 | ||
| BC112172.1 | - | 2 | 193 | 8327640 | ||
| W77994.1 | - | 1 | 128 | |||

SPRR4
Genomic Map