siRNA Details
| Gene Symbol: FAM71D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173526.3 | NP_775797.2 | 9 | 1751 | ||
| XM_011536500.1 | XP_011534802.1 | 9 | 1668 | |||
| XM_011536502.1 | XP_011534804.1 | 9 | 1650 | |||
| XM_011536503.2 | XP_011534805.1 | 8 | 2692 | |||
| GenBank mRNA | AK093449.1 | - | 9 | 1715 | ||
| BC050401.1 | - | 10 | 1617 | 5745029 | ||

FAM71D
Genomic Map