siRNA Details
| Gene Symbol: KCTD20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173562.4 | NP_775833.2 | 2, 3 | 561 | ||
| XM_005248940.4 | XP_005248997.1 | 3, 4 | 416 | |||
| XM_006715023.2 | XP_006715086.1 | 2, 3 | 334 | |||
| XM_011514398.2 | XP_011512700.1 | 3, 4 | 412 | |||
| GenBank mRNA | AK304377.1 | - | 2, 3 | 282 | ||
| AL832381.1 | - | 1, 2 | 73 | |||
| AY305862.1 | - | 2, 3 | 561 | |||

KCTD20
Genomic Map