siRNA Details
| Gene Symbol: FAM217A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173563.2 | NP_775834.2 | 7 | 1390 | ||
| XM_005248951.3 | XP_005249008.1 | 6 | 1310 | |||
| XM_006715025.2 | XP_006715088.1 | 5 | 1116 | |||
| XM_006715026.3 | XP_006715089.1 | 5 | 1521 | |||
| XM_011514413.2 | XP_011512715.1 | 6 | 1436 | |||
| XM_011514414.2 | XP_011512716.1 | 4 | 1337 | |||
| XM_011514415.1 | XP_011512717.1 | 4 | 1157 | |||
| XM_011514416.2 | XP_011512718.1 | 4 | 1509 | |||
| XM_011514417.1 | XP_011512719.1 | 5 | 974 | |||
| XM_017010480.1 | XP_016865969.1 | 6 | 1609 | |||
| XM_017010481.1 | XP_016865970.1 | 5 | 1594 | |||
| XM_017010482.1 | XP_016865971.1 | 6 | 1407 | |||
| GenBank mRNA | AK097426.1 | - | 5 | 1394 | ||
| BC039349.1 | - | 7 | 1324 | 5269651 | ||

FAM217A
Genomic Map