siRNA Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079519.1 | NP_001072987.1 | 7 | 1900 | ||
| NM_001289022.1 | NP_001275951.1 | 6 | 1623 | |||
| NM_173607.3 | NP_775878.2 | 5 | 1570 | |||
| GenBank mRNA | AK096173.1 | - | 6 | 1506 | ||
| AL833508.1 | - | 5 | 1484 | |||
| BX640884.1 | - | 5 | 1573 | |||
| BX640984.1 | - | 5 | 1521 | |||
| CR749796.1 | - | 5 | 1570 | |||

FAM177A1
Genomic Map