siRNA Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079519.1 | NP_001072987.1 | 7 | 2841 | ||
| NM_001289022.1 | NP_001275951.1 | 6 | 2564 | |||
| NM_173607.3 | NP_775878.2 | 5 | 2511 | |||
| GenBank mRNA | AK096173.1 | - | 6 | 2447 | ||
| AL833508.1 | - | 5 | 2425 | |||
| BC040538.1 | - | 3379 | ||||
| BX640884.1 | - | 5 | 2514 | |||
| BX640984.1 | - | 5 | 2462 | |||
| CD678775.1 | - | 1 | 419 | |||
| CR749796.1 | - | 5 | 2509 | |||

FAM177A1
Genomic Map