siRNA Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173854.5 | NP_776253.3 | 11 | 4294 | ||
| XM_005245069.2 | XP_005245126.1 | 11 | 3970 | |||
| GenBank mRNA | AK092123.1 | - | 1 | 1048 | ||
| AK125075.1 | - | 10 | 3055 | |||
| AK128069.1 | - | 11 | 3446 | |||
| AL831974.1 | - | 11 | 4130 | |||
| AL832001.1 | - | 11 | 4130 | |||
| AL832362.1 | - | 11 | 4130 | |||
| BC136303.1 | - | 10 | 3775 | 9020290 | ||
| BX641054.1 | - | 11 | 4134 | |||
| BX648979.1 | - | 11 | 4168 | |||

SLC41A1
Genomic Map